missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-DR Antibody (L243) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 29 publications
£188.00
Specifications
| Antigen | HLA-DR |
|---|---|
| Clone | L243 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, Flow Cytometry < 0.5 μg/106 cells, ELISA reported in scientific literature (PMID 11070170), Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 7835916), Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen 1:10 - 1:500, Functional reported in scientific literature (PMID 17900279), In vitro assay reported in scientific literature (PMID 19620786), Electron Microscopy reported in scientific literature (PMID 21660937), Flow (Cell Surface), CyTOF-ready, Block/Neutralize reported in scientific literature (PMID 12874328), Dual RNAscope ISH-IHC |
| Applications | Western Blot |
Description
HLA-DR Monoclonal specifically detects HLA-DR in Human, Canine, Baboon, Primate samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Functional, In vitro assay, Electron Microscopy, Flow (Cell Surface), CyTOF-ready, Block/Neutralize, Dual RNAscope ISH-IHC.Specifications
| HLA-DR | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cell Biology, Cellular Markers, Diabetes Research, Immunology, Major Histocompatibility Complexes | |
| P01903 | |
| 3122 | |
| Human lymphoblastoid cell line (RPMI 8866). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| 28 kDa |
| L243 | |
| Western Blot 1:100 - 1:2000, Flow Cytometry < 0.5 μg/106 cells, ELISA reported in scientific literature (PMID 11070170), Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 7835916), Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen 1:10 - 1:500, Functional reported in scientific literature (PMID 17900279), In vitro assay reported in scientific literature (PMID 19620786), Electron Microscopy reported in scientific literature (PMID 21660937), Flow (Cell Surface), CyTOF-ready, Block/Neutralize reported in scientific literature (PMID 12874328), Dual RNAscope ISH-IHC | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| FLJ51114, histocompatibility antigen HLA-DR alpha, HLA class II histocompatibility antigen, DR alpha chain, HLA-DRA1, major histocompatibility complex, class II, DR alpha, MHC cell surface glycoprotein, MHC class II antigen DRA, MLRW | |
| HLA-DRA | |
| IgG2a κ | |
| Protein A purified | |
| Human. Predicted cross-reactivity with Chimpanzee, Baboon, Cynomolgus, Rhesus, Marmoset, Tamarin, Squirrel Monkey and Canine. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title