missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-DR Antibody (L243) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 29 publications
Brand: Novus Biologicals NB100-77855SS
This item is not returnable.
View return policy
Description
HLA-DR Monoclonal specifically detects HLA-DR in Human, Canine, Baboon, Primate samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Functional, In vitro assay, Electron Microscopy, Flow (Cell Surface), CyTOF-ready, Block/Neutralize, Dual RNAscope ISH-IHC.
Specifications
| HLA-DR | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:2000, Flow Cytometry < 0.5 μg/106 cells, ELISA reported in scientific literature (PMID 11070170), Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 7835916), Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen 1:10 - 1:500, Functional reported in scientific literature (PMID 17900279), In vitro assay reported in scientific literature (PMID 19620786), Electron Microscopy reported in scientific literature (PMID 21660937), Flow (Cell Surface), CyTOF-ready, Block/Neutralize reported in scientific literature (PMID 12874328), Dual RNAscope ISH-IHC | |
| FLJ51114, histocompatibility antigen HLA-DR alpha, HLA class II histocompatibility antigen, DR alpha chain, HLA-DRA1, major histocompatibility complex, class II, DR alpha, MHC cell surface glycoprotein, MHC class II antigen DRA, MLRW | |
| Mouse | |
| 28 kDa | |
| 0.025 mL | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cell Biology, Cellular Markers, Diabetes Research, Immunology, Major Histocompatibility Complexes | |
| 3122 | |
| Human | |
| Purified |
| Western Blot | |
| L243 | |
| Unconjugated | |
| P01903 | |
| HLA-DRA | |
| Human lymphoblastoid cell line (RPMI 8866). | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human. Predicted cross-reactivity with Chimpanzee, Baboon, Cynomolgus, Rhesus, Marmoset, Tamarin, Squirrel Monkey and Canine. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction