missing translation for 'onlineSavingsMsg'
Learn More

HLA DOA Antibody [Unconjugated], Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP3-43684-25ul

 View more versions of this product

Product Code. 30279472

  • £321.00 / 25µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

HLA DOA Polyclonal antibody specifically detects HLA DOA in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HLA DOA
Polyclonal
Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
HLA class II histocompatibility antigen, DO alpha chain, HLA-D0-alpha, HLA-DNAHLADZ, HLA-DZA, lymphocyte antigen, major histocompatibility complex, class II, DN alpha, major histocompatibility complex, class II, DO alpha, MHC class II antigen DOA, MHC DN-alpha, MHC DZ alpha
This antibody was developed against Recombinant Protein corresponding to amino acids: ATKADHMGSYGPAFYQSYGASGQFTHEFDEEQLFSVDLKKSEAVWRLPEF
25 μL
Asthma, Diabetes Research, Immunology
3111
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
PBS, pH 7.2, 40% glycerol
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.