missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA DMA Antibody (6B12NB) - Azide Free, Novus Biologicals™
Mouse Monoclonal Antibody
£143.00 - £378.00
Specifications
| Antigen | HLA DMA |
|---|---|
| Clone | 6B12NB |
| Concentration | 1.0mg/mL |
| Dilution | Western Blot 2 ug/ml, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 10-20 ug/ml, Immunohistochemistry-Paraffin 1:200, Flow (Cell Surface) 2.5 ug/million cells |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18189288
|
Novus Biologicals
NBP2-44304SS |
0.025 mg |
£143.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18118888
|
Novus Biologicals
NBP2-44304 |
0.1 mg |
£378.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ensure accurate, reproducible results in Western Blot, Flow (Cell Surface), Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)
HLA DMA Monoclonal specifically detects HLA DMA in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Cell Surface).Specifications
| HLA DMA | |
| 1.0mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Asthma, Diabetes Research, Immunology | |
| PBS with No Preservative | |
| 3108 | |
| Partial recombinant human HLA DMA protein (between amino acids 10-300) [Entrez NP_006111] | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| 6B12NB | |
| Western Blot 2 ug/ml, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 10-20 ug/ml, Immunohistochemistry-Paraffin 1:200, Flow (Cell Surface) 2.5 ug/million cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| class II histocompatibility antigen, M alpha chain, D6S222E, DMA, HLA class II histocompatibility antigen, DM alpha chain, major histocompatibility complex, class II, DM alpha, MHC class II antigen DMA, Really interesting new gene 6 protein, RING6HLADM | |
| HLA-DMA | |
| IgG1 κ | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title