missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA DMA Antibody (6B12NB) - Azide Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-44304SS
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot, Flow (Cell Surface), Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)
HLA DMA Monoclonal specifically detects HLA DMA in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Cell Surface).
Specifications
| HLA DMA | |
| Monoclonal | |
| 1.0mg/mL | |
| Western Blot 2 ug/ml, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 10-20 ug/ml, Immunohistochemistry-Paraffin 1:200, Flow (Cell Surface) 2.5 ug/million cells | |
| class II histocompatibility antigen, M alpha chain, D6S222E, DMA, HLA class II histocompatibility antigen, DM alpha chain, major histocompatibility complex, class II, DM alpha, MHC class II antigen DMA, Really interesting new gene 6 protein, RING6HLADM | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 6B12NB | |
| Unconjugated | |
| PBS with No Preservative | |
| HLA-DMA | |
| Partial recombinant human HLA DMA protein (between amino acids 10-300) [Entrez NP_006111] | |
| 0.025 mg | |
| Asthma, Diabetes Research, Immunology | |
| 3108 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction