missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HK1 Recombinant Rabbit Monoclonal Antibody (23GB870), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | HK1 |
|---|---|
| Clone | 23GB870 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30087991
![]() |
Invitrogen™
MA554162 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089623
|
Invitrogen™
MA554163 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HK1 Recombinant Monoclonal antibody specifically detects HK1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| HK1 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P05708, P17710, P19367 | |
| 15275, 25058, 3098 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB870 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| BB404130; brain form hexokinase; dea; downeast anemia; glycolytic enzyme; HEXOKIN; hexokinase; hexokinase 1; hexokinase I; hexokinase IR; hexokinase type I; Hexokinase, tumor isozyme; hexokinase-1; Hexokinase-A; HK; HK I; HK1; Hk-1; Hk1-s; HK1-ta; HK1-tb; HK1-tc; HKD; HKI; HMSNR; HXK1; mHk1-s | |
| A synthetic peptide of human Hexokinase 1 (100-156AA). | |
| Primary | |
| -20°C | |
| HK1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title