missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HK1 Recombinant Rabbit Monoclonal Antibody (23GB870), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA554162
This item is not returnable.
View return policy
Description
This gene encodes a member of the ARHGAP family of Rho/Rac/Cdc42-like GTPase activating proteins. The encoded protein interacts with the Ras-related protein Gem through its N-terminal domain. Separately, it interacts with RhoA through a RhoGAP domain, and stimulates RhoA-dependent GTPase activity. Alternatively spliced variants that encode different protein isoforms have been described but their full-length nature has not been determined.
Specifications
| HK1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| HK1 | |
| BB404130; brain form hexokinase; dea; downeast anemia; glycolytic enzyme; HEXOKIN; hexokinase; hexokinase 1; hexokinase I; hexokinase IR; hexokinase type I; Hexokinase, tumor isozyme; hexokinase-1; Hexokinase-A; HK; HK I; HK1; Hk-1; Hk1-s; HK1-ta; HK1-tb; HK1-tc; HKD; HKI; HMSNR; HXK1; mHk1-s | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 15275, 25058, 3098 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB870 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P05708, P17710, P19367 | |
| HK1 | |
| A synthetic peptide of human Hexokinase 1 (100-156AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction