Learn More
Cytiva HiTrap™ Protein L Prepacked Columns
For capture of antibodies and antibody fragments such as Fab fragments, scFv, and Dabs
Brand: Cytiva 29048665
Code : X3
Additional Details : CAS Number : 64-17-5 Weight : 0.09800kg
Description
- Prepacked with Capto L affinity chromatography medium (resin)
- Polypropylene column are easily connected in series for increased productivity
- Easy screening and convenient process development
- For use with syringe, peristaltic pump, or chromatography system
- Maximum pressure: 5 bar
- 1 and 5mL columns
BioProcess Media
- Ligand: Recombinant protein L (E. coli), mammalian free
- Matrix: Rigid, highly cross-linked agarose
- Average particle size: 85μm
- pH stability working range: 2 to 10
1mL columns
- Column I.D.: 7mm
- Flow velocity: <500cm/h3
5mL columns
- Column I.D.: 16mm
- Flow velocity: <500cm/h4
Specifications
HiTrap Protein L column | |
0.27 in. | |
1 x 1mL | |
20mm | |
Polypropylene | |
5 bar | |
85μm | |
2° to 8°C | |
Affinity chromato graphy medium (Resin) for capture of Antibodies and Antibody fragments such as Fab fragments, scFv and Dabs |
1mL | |
7mm | |
0.78 in. | |
<500cm/hr. | |
Recombinant Protein L (E. coli), mammalian free | |
2 to 10 | |
Highly Cross-linked Agarose | |
Affinity Column |
Safety and Handling
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol