missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ Protein L Prepacked Columns

Product Code. 12671206 Shop All Cytiva Products
Click to view available options
Quantity:
1 x 1mL
1 x 5mL
5 x 1mL
5 x 5mL
Diameter (English) Inner:
0.27 in.
0.62 in.
Unit Size:
Each
Pack of 5
This item is not returnable. View return policy

Product Code. 12671206

Brand: Cytiva 17547815

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For capture of antibodies and antibody fragments such as Fab fragments, scFv, and Dabs

  • Prepacked with Capto L affinity chromatography medium (resin)
  • Polypropylene column are easily connected in series for increased productivity
  • Easy screening and convenient process development
  • For use with syringe, peristaltic pump, or chromatography system
  • Maximum pressure: 5 bar
  • 1 and 5mL columns

BioProcess Media
  • Ligand: Recombinant protein L (E. coli), mammalian free
  • Matrix: Rigid, highly cross-linked agarose
  • Average particle size: 85μm
  • pH stability working range: 2 to 10

1mL columns

  • Column I.D.: 7mm
  • Flow velocity: <500cm/h3

5mL columns
  • Column I.D.: 16mm
  • Flow velocity: <500cm/h4

TRUSTED_SUSTAINABILITY

Specifications

Product Type HiTrap Protein L column
Bed Volume 5mL
Diameter (English) Inner 0.62 in.
Diameter (Metric) Inner 16mm
Quantity 1 x 5mL
Height (English) 0.78 in.
Height (Metric) 20mm
Flow Rate <500 cm/hr.
Material Polypropylene
Ligand Type Recombinant Protein L (E. coli), mammalian free
Max. Pressure 5 bar
pH Range 2 to 10
Particle Size 85μm
Support Type Highly Cross-linked Agarose
Content And Storage 2° to 8°C
Column Type Affinity Column
For Use With (Application) Affinity chromato graphy medium (Resin) for capture of Antibodies and Antibody fragments such as Fab fragments, scFv and Dabs
Show More Show Less
compliance-icons
Product Identifier
  • HiTrap Protein L, 1 x 5 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ethanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.