missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ Chelating HP Columns

Product Code. 10773017 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 x 5mL
5 x 1mL
5 x 5mL
100 x 5 mL
Unit Size:
Each
Pack of 100
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10773017 1 x 5mL Each
11850073 100 x 5 mL Pack of 100
10605575 5 x 1mL Pack of 5
11594895 5 x 5mL Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10773017 Supplier Cytiva Supplier No. 17040901

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Optimize histidine-tagged protein purifications when nickel ions are not the best choice. Cytiva Life Sciences™ HiTrap™ Chelating HP Columns is prepacked with Chelating Sepharose™ High Performance.

  • Uses different kinds of metal ions
  • Binding capacity ∽12mg histidine-tagged protein/mL medium
  • Metal ion capacity ∽23μmol Cu2+/mL medium
  • Prepacked with Chelating Sepharose High Performance media
  • Iminoacetic acid coupled to a 6% agarose matrix by ether linkage

TRUSTED_SUSTAINABILITY

Specifications

Buffer 6M Guanidine Hydrochloride, 8M Urea
Product Type HiTrap chelating HP column
Bed Volume 5mL
Diameter (English) Inner 0.62 in.
Diameter (Metric) Inner 16mm
Quantity 1 x 5mL
Height (English) 0.98 in.
Height (Metric) 25mm
Flow Rate <20 mL/min.
Material Polypropylene
Ligand Type Iminodiacetic Acid
Max. Pressure 3 bar
pH Range 3 to 13
Particle Size 34μm
Support Type 6% Highly Cross-linked Agarose
Content And Storage 4° to 30°C
Column Type Affinity Column
For Use With (Application) IMAC and subsequent purification of Polyhistidine tagged Proteins
Show More Show Less
compliance-icons
Product Identifier
  • HiTrap Chelating HP 5 ml, 1 x 5 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.