Learn More
Cytiva HiTrap Capto Phenyl (High Sub)
Capto Phenyl (High sub) and Capto Butyl are hydrophobic interaction chromatography (HIC) resins used in the capture and intermediate stages of protein purification
Brand: Cytiva 17545108
Additional Details : Weight : 0.08000kg
Description
- Hydrophobic interaction chromatography (HIC) resins
- Used in the capture and intermediate stages of protein purification
Specifications
1 mL | |
< 600 cm/h | |
5 x 1 mL | |
4 to 30 °C, 20% Ethanol |
7 mm | |
Hydrophobic Interaction Chromatography | |
25 mm |
Safety and Handling
- HiTrap Capto Phenyl (high sub) 1 ml, 5 x 1 ml
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.