missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ Capto™ Core 700 Columns

Product Code. 12643860 Shop All Cytiva Products
Click to view available options
Quantity:
5 x 1mL
Unit Size:
Pack of 5
This item is not returnable. View return policy

Product Code. 12643860

Brand: Cytiva 17548151

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

HiTrap Capto Core 700 is prepacked with Capto Core 700, a multimodal BioProcess medium for intermediate purification and polishing of viruses and other large biomolecules.

Capto Core 700 is a novel, layered bead chromatography medium consisting of a porous outer layer and ligand-activated core.

  • High capacity and productivity in flowthrough mode.
  • Wide operational window of pH and conductivity.
  • Prepacked HiTrap columns for easy screening and convenient process development, as well as small scale purification.
  • The octylamine ligands found in the core of the beads are multimodal, being both hydrophobic and positively charged.
  • These internalized ligands bind impurities strongly over a wide range of pH and salt concentration.
  • The pores of the bead outer layer prevent large targets (Mr > 700 000) from binding to the ligands while smaller impurities can enter freely into the beads where they are captured.

TRUSTED_SUSTAINABILITY

Specifications

Buffer 1M NaOH, 6M Guanidine Hydrochloride, 30% Isopropanol and 70% Ethanol
Product Type HiTrap Capto Core 700 multimodal column
Bed Volume 1mL
Diameter (English) Inner 0.27 in.
Diameter (Metric) Inner 7mm
Quantity 5 x 1mL
Flow Rate <500cm/hr.
Material Polypropylene
Ligand Type Octylamine
Max. Pressure 5 bar
pH Range 2 to 14
Particle Size 90μm
Support Type Highly Cross-linked Agarose
Content And Storage 4° to 30°C
Column Type Multimodal Column
For Use With (Application) Intermediate purification and polishing of viruses and other large biomolecules
Show More Show Less
compliance-icons
Product Identifier
  • HiTrap Capto Core 700, 5 x 1 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.