missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ Capto™ Adhere Columns

Product Code. 10635935 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
5 x 1mL
5 x 5mL
Diameter (English) Inner:
0.30 in.
0.62 in.
Flow Rate:
<20 mL/min.
<4 mL/min.
Max. Pressure:
1.5 bar
44 bar
Unit Size:
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Diameter (English) Inner Flow Rate Max. Pressure unitSize
10635935 5 x 1mL 0.30 in. <4 mL/min. 1.5 bar Pack of 5
11505135 5 x 5mL 0.62 in. <20 mL/min. 44 bar Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10635935 Supplier Cytiva Supplier No. 28405844

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Designed to remove contaminants in post-protein A purification

  • Prepacked with Capto adhere media, a strong anion exchanger with multimodal functionality
  • High capacity and productivity in flow-through mode: 0.09 to 0.12mmol Cl-/mL medium ionic capacity
  • Wide operational window of pH and conductivity: working pH range 3 to 12
  • Two-step process reduces time spent and operating costs

TRUSTED_SUSTAINABILITY

Specifications

Buffer Aqueous Buffer
Product Type HiTrap Capto adhere column
Bed Volume 1mL
Diameter (English) Inner 0.30 in.
Diameter (Metric) Inner 7mm
Quantity 5 x 1mL
Height (English) 0.98 in.
Height (Metric) 25mm
Flow Rate <4 mL/min.
Material Polypropylene
Ligand Type N-benzyl-n-methyl Ethanolamine
Max. Pressure 1.5 bar
pH Range 3 to 12
Particle Size 75μm
Support Type Highly Cross-linked Agarose
Content And Storage 4° to 30°C
Column Type Affinity Column
For Use With (Application) Easy screening and convenient process development
Show More Show Less
compliance-icons
Product Identifier
  • HiTrap Capto adhere 1 ml, 5 x 1 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.