missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX HitHunter™ cAMP Assay for Biologics
Click to view available options
Quantity:
200/800 dp (96-well/384-well)
Unit Size:
1 set
Description
This detection kit is a robust, sensitive and easy-to-use assay to determine GPCR activity through detection of intracellular cAMP levels. The HitHunter cAMP assay is intended for use with cells expressing Gs or Gi coupled GPCRs. This product uses EFC technology, and contains ED-tagged cAMP, anti-cAMP antibody, and pure EA. Upon application to a cell which contains endogenous, untagged cAMP, the anti-cAMP antibody will be sequestered by binding to this untagged cAMP, allowing for EFC between the tagged cAMP and soluble EA. This will produce a proportional level of chemiluminescent signal. This kit format is intended for Biologics, including crude preps, antibodies, and more, and is written for a 96-well plate. This kit may be used with Eurofins DiscoverX cAMP cell lines as well as cell lines from other sources. This detection kit contains reagents for assaying 2 plates (96-well).
Specifications
Specifications
| Assay Entry | GPCR cAMP |
| For Use With (Application) | Detection Kits |
| Quantity | 200/800 dp (96-well/384-well) |
| Target Entry | cAMP |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction