missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HisTrap™ FF Crude Prepacked Columns

Product Code. 10685355 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
5 x 5 mL
60 mL
100 x 1 mL
100 x 5 mL
Flow Rate:
<20 mL/min.
<4 mL/min.
Unit Size:
Pack of 100
Pack of 5
Product Code. Quantity Flow Rate unitSize
10685355 60 mL <4 mL/min. Pack of 5
11723219 5 x 5 mL <20 mL/min. Pack of 5
11515674 100 x 1 mL <4 mL/min. Pack of 100
11800083 100 x 5 mL <20 mL/min. Pack of 100
4 options
This item is not returnable. View return policy

Product Code. 10685355

Brand: Cytiva 11000458

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Purify histidine-tagged proteins directly from homogenized, unclarified cell lysates. Cytiva Life Sciences™ HisTrap™ FF Crude Prepacked Columns offer the convenience of simple, one-step purification of histidine-tagged proteins by immobilized metal ion affinity chromatography (IMAC).

  • Prepacked with Ni Sepharose™ 6 Fast Flow media
  • Binding capacity approximately 40mg/mL medium with negligible Ni2+ leakage
  • Maximum pressure: 3 bar
  • Column material: polypropylene
  • Compatible with wide variety of reducing agents, detergents, denaturants, and other additives
  • 1 and 5mL columns

Media
  • Matrix: 6% cross-linked agarose
  • Average particle size: 90μm
  • pH stability working range: 3 to 12

1mL columns

  • Bed dimensions: 7 x 25mm
  • Flow velocity: <4mL/min.

5mL columns
  • Bed dimensions: 16 x 25mm
  • Flow velocity: <20mL/min.

TRUSTED_SUSTAINABILITY

Specifications

Bed Volume 1 mL
Diameter (English) Inner 0.2 in.
Diameter (Metric) Inner 7 mm
Description HisTrap FF Crude columns are high binding capacity IMAC columns for his-tagged protein purification when scalability is needed.
Flow Rate <4 mL/min.
Kit Contents Sodium Phosphate-buffered Saline, pH 7.4 and 0.5X concentration of proprietary Y-PER reagent additives
Material Polypropylene
pH Range 3 to 12
Support Type 6% Cross-linked Agarose
Content And Storage 4°C to 30°C
Max. Operating Pressure 5 bar (0.5 MPa, 70 psi)
Particle Size 45 to 165 μm
Quantity 60 mL
For Use With (Application) Intended for research use only, and shall not be used in any clinical or in vitro procedures for diagnostic purposes
Type Pre-packed Column
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : ethanol, Nickel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.