Learn More
Cytiva HisTrap™ FF Columns: 16 x 25mm
Use for optimized purification of histidine-tagged proteins directly from homogenized, unclarified cells by immobilized metal ion affinity chromatography (IMAC). Cytiva Life Sciences™ HisTrap™ FF Columns: 16 x 25mm is a ready-to-use column, prepacked with precharged Ni Sepharose™ 6 Fast Flow.
Brand: Cytiva 17-5319-02
Code : GX
Additional Details : CAS Number : 79-06-1 Weight : 1,134.00000kg
Description
- Prepacked with precharged Ni Sepharose™ 6 Fast Flow media
- Column material: polypropylene
- Bed dimensions: 16 × 25mm
- Flow velocity: <20mL/min.
- Protein binding capacity of ∽40mg/mL media with negligible nickel ion leakage
- Compatible with wide range of reducing agents, detergents, denaturants, and other additives
- Simple operation with syringe, perstaltic pump, or chromatographic system
- Columns may be connected in series to increase capacity
Medium
- Matrix: 6% cross-linked agarose
- Average particle size: 90μm
- pH stability working range: 3 to 12
Specifications
5 mL | |
16 mm | |
<20 mL/min. | |
3 to 12 | |
4°C to 30°C, 20% Ethanol | |
100 x 1 mL |
0.6 in. | |
HisTrap FF is a ready-to-use column, prepacked with precharged Ni Sepharose 6 Fast Flow for preparative purification of histidine-tagged recombinant proteins by immobilized metal ion affinity chromatography (IMAC). | |
Polypropylene | |
6% Cross-linked Agarose | |
5 bar (0.5 MPa, 70 psi) | |
Pre-packed Column |
Safety and Handling
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : ethanol, Nickel