missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H3, p Ser10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£259.00 - £423.00
Specifications
| Applications | Immunohistochemistry, Immunocytochemistry |
|---|---|
| Classification | Polyclonal |
| Gene Alias | H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, H3S10p, HIST2H3C, histone H3.3, MGC87782, MGC87783 |
| Gene Symbols | H3C14 |
| Content And Storage | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18775433
|
Novus Biologicals
NB21-1091SS |
£259.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
|
18791214
|
Novus Biologicals
NB21-1091 |
0.05 mg |
£423.00
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Histone H3 Polyclonal specifically detects Histone H3 in Human, Mouse, Rat, C. elegans samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Dot Blot, Single Cell Western.Specifications
| Immunohistochemistry, Immunocytochemistry | |
| H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, H3S10p, HIST2H3C, histone H3.3, MGC87782, MGC87783 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Polyclonal | |
| H3C14 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title