Learn More
Histone H3, p Ser10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB21-1091
This item is not returnable.
View return policy
Description
Histone H3 Polyclonal specifically detects Histone H3 in Human, Mouse, Rat, C. elegans samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Dot Blot, Single Cell Western.
Specifications
| Histone H3 (p Ser10) | |
| Polyclonal | |
| RUO | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| 0.05 mg | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |