missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H3, ac Lys4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£234.00 - £399.00
Specifications
| Isotype | IgG |
|---|---|
| Content And Storage | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18780254
|
Novus Biologicals
NB21-1024SS |
£234.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
|
18264770
|
Novus Biologicals
NB21-1024 |
0.05 mg |
£399.00
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Histone H3 Polyclonal specifically detects Histone H3 in Human, Mouse, Rat, C. elegans samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunocytochemistry/Immunofluorescence, Dot Blot.Specifications
| IgG | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title