Learn More
Histone H3, ac Lys4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand:  Novus Biologicals NB21-1024SS
 View more versions of this product
Product Code. 18780254
  • £234.00 / 25µg
Description
Histone H3 Polyclonal specifically detects Histone H3 in Human, Mouse, Rat, C. elegans samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunocytochemistry/Immunofluorescence, Dot Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.