Learn More
Histone H3, ac Lys4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand:  Novus Biologicals NB21-1024
 View more versions of this product
Product Code. 18264770
  • £399.00 / 0.05mg
Description
Histone H3 Polyclonal specifically detects Histone H3 in Human, Mouse, Rat, C. elegans samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunocytochemistry/Immunofluorescence, Dot Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Histone H3 (ac Lys4)
Polyclonal
Unconjugated
PBS and 30% Glycerol with 0.05% Sodium Azide
H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, H3K4ac, HIST2H3C, histone H3.3, MGC87782, MGC87783
Rabbit
15 kDa
0.05 mg
Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls
126961
Human, Mouse, Rat, C. elegans
IgG
Western Blot, Immunocytochemistry, Immunofluorescence, Dot Blot, ChIP Assay
1.15 mg/mL
Western Blot 1:500, Immunocytochemistry/Immunofluorescence 1:1000, Dot Blot 1:1000, Chromatin Immunoprecipitation (ChIP) 2-5 ug per million cells
Q71DI3, P84228
H3C14
This Histone H3 [ac Lys4] antibody was raised against synthetic acetylated peptide surrounding Lysine 4 of human Histone H3.2 [Swiss Prot Q71DI3].
Affinity Purified
RUO
Primary
Human, mouse, and S. cerevisiae. Predicted to react with many species including rat, chicken, Xenopus, Drosophila, and plant based on 100% sequence homology.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.