missing translation for 'onlineSavingsMsg'
Learn More

Histone H2B, ubiquitylated Lys123 Antibody (1B3F12/A9) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication

£169.00 - £340.00

Specifications

Antigen Histone H2B
Clone 1B3F12/A9
Concentration 1.0 mg/mL
Dilution Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence, Chromatin Immunoprecipitation (ChIP) 1:10-1:500
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, ChIP Assay
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18280830
View Documents
Novus Biologicals
NBP2-54770
0.1 mL
£340.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18667197
View Documents
Novus Biologicals
NBP2-54770-0.025ml
0.025 mL
£169.00
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Histone H2B Monoclonal specifically detects Histone H2B in Human, S. pombe, Yeast samples. It is validated for Western Blot, Chromatin Immunoprecipitation, ELISA, Immunocytochemistry/Immunofluorescence, Chromatin Immunoprecipitation (ChIP).
Specifications

Specifications

Histone H2B
1.0 mg/mL
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, ChIP Assay
Unconjugated
Mouse
Chromatin Research, Epigenetics, Histones and Modified Histones
PBS with 0.02% Sodium Azide
440689
Ub-polypeptide antigen based on amino acids 115 - 130 of yeast Histone H2B
Primary
23 kDa
1B3F12/A9
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence, Chromatin Immunoprecipitation (ChIP) 1:10-1:500
Monoclonal
Purified
RUO
Human, S. pombe, Yeast
FLJ35099, FLJ56780, FLJ56787, histone 2, H2bf, histone cluster 2, H2bf, histone H2B type 2-F, MGC131639
HIST2H2BF
IgG1
Protein A purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.