missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H2B, ubiquitylated Lys123 Antibody (1B3F12/A9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-54770-0.025ml
This item is not returnable.
View return policy
Description
Histone H2B Monoclonal specifically detects Histone H2B in Human, S. pombe, Yeast samples. It is validated for Western Blot, Chromatin Immunoprecipitation, ELISA, Immunocytochemistry/Immunofluorescence, Chromatin Immunoprecipitation (ChIP).
Specifikationer
| Histone H2B | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 | |
| FLJ35099, FLJ56780, FLJ56787, histone 2, H2bf, histone cluster 2, H2bf, histone H2B type 2-F, MGC131639 | |
| Mouse | |
| 23 kDa | |
| 0.025 mL | |
| Chromatin Research, Epigenetics, Histones and Modified Histones | |
| 440689 | |
| Human, S. pombe, Yeast | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, ChIP Assay | |
| 1B3F12/A9 | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| HIST2H2BF | |
| Ub-polypeptide antigen based on amino acids 115 - 130 of yeast Histone H2B | |
| Protein A purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering