missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H2AY/macroH2A.1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-53003
This item is not returnable.
View return policy
Description
Histone H2AY/macroH2A.1 Polyclonal specifically detects Histone H2AY/macroH2A.1 in Human samples. It is validated for Western Blot.
Specifications
| Histone H2AY/macroH2A.1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| core histone macro-H2A.1, H2A histone family, member Y, H2A.y, H2A/y, H2AF12M, H2AFJ, Histone H2A.y, Histone macroH2A1, histone macroH2A1.1, histone macroH2A1.2, MACROH2A1, MACROH2A1.1, macroH2A1.2, Medulloblastoma antigen MU-MB-50.205, mH2A1 | |
| Rabbit | |
| 39 kDa | |
| 100 μL | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| O75367 | |
| H2AFY | |
| Synthetic peptides corresponding to H2AFY(H2A histone family, member Y) The peptide sequence was selected from the N terminal of H2AFY. Peptide sequence HPKYRIGVGAPVYMAAVLEYLTAEILELAGNAARDNKKGRVTPRHILLAV. | |
| Affinity purified | |
| RUO | |
| 9555 | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Yeast, Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction