missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H2AX Antibody (S02-3C8), Novus Biologicals™
Rabbit Monoclonal Antibody
£328.00 - £501.00
Specifications
| Antigen | Histone H2AX |
|---|---|
| Clone | S02-3C8 |
| Concentration | 0.3 mg/mL |
| Dilution | Western Blot, Immunohistochemistry 1:50, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18672225
|
Novus Biologicals
NBP3-20068-100ul |
100 μg |
£501.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18673395
|
Novus Biologicals
NBP3-20068-50ul |
50 μg |
£328.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Histone H2AX Monoclonal antibody specifically detects Histone H2AX in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| Histone H2AX | |
| 0.3 mg/mL | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Mitotic Regulators, Phospho Specific | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 3014 | |
| IgG | |
| Affinity purified |
| S02-3C8 | |
| Western Blot, Immunohistochemistry 1:50, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| H2A.X, H2A/X, H2AFX | |
| A synthetic peptide of human Histone H2AX (Uniprot # P16104) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title