missing translation for 'onlineSavingsMsg'
Learn More

Histone H2AX Antibody (S02-3C8), Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals NBP3-20068-100ul

 View more versions of this product

Product Code. 18672225

  • £501.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Histone H2AX Monoclonal antibody specifically detects Histone H2AX in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Histone H2AX
Monoclonal
0.3 mg/mL
Western Blot, Immunohistochemistry 1:50, Immunohistochemistry-Paraffin
H2A.X, H2A/X, H2AFX
A synthetic peptide of human Histone H2AX (Uniprot # P16104)
100 μg
Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Mitotic Regulators, Phospho Specific
3014
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
S02-3C8
Unconjugated
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.