missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H1T Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£149.00 - £290.00
Specifications
| Antigen | Histone H1T |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18348665
|
Novus Biologicals
NBP3-15574-20UL |
20 μg |
£149.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18397356
|
Novus Biologicals
NBP3-15574-100UL |
100 μg |
£290.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Histone H1T Polyclonal antibody specifically detects Histone H1T in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
| Histone H1T | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| dJ221C16.2, H1 histone family, member T (testis-specific), H1FTMGC163222, H1t, histone 1, H1t, histone cluster 1, H1t, histone H1t, Testicular H1 histone | |
| A synthetic peptide corresponding to a sequence within amino acids 150-209 of mouse Histone H1T (NP_034507.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Epigenetics | |
| PBS, 50% glycerol, pH7.3 | |
| 3010 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title