missing translation for 'onlineSavingsMsg'
Learn More

Histone H1T Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™

Product Code. 18397356 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18397356 100 μg 100µL
18348665 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18397356 Supplier Novus Biologicals Supplier No. NBP315574100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Histone H1T Polyclonal antibody specifically detects Histone H1T in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histone H1T
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias dJ221C16.2, H1 histone family, member T (testis-specific), H1FTMGC163222, H1t, histone 1, H1t, histone cluster 1, H1t, histone H1t, Testicular H1 histone
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 150-209 of mouse Histone H1T (NP_034507.2).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 3010
Target Species Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.