missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H1 Mouse, Clone: r1415-1, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 3005-MSM7-P0
Additional Details : Weight : 0.01000kg
Specifications
Histone H1 | |
Nuclear | |
r1415-1 | |
Unconjugated | |
NP_005309 | |
Mouse | |
∽30kDa | |
Protein A/G Purified | |
RUO | |
3005 | |
IgG2a κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with or without 0.05% BSA and 0.05% azide. | |
H1 | |
Nuclei of human leukemia biopsy cells | |
HeLa, A-431, LNCap or Jurkat cells. Breast carcinoma. | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |