missing translation for 'onlineSavingsMsg'
Learn More

Histone H1 Mouse, Clone: r1415-1, enQuireBio™

Mouse Monoclonal Antibody

£181.83 - £395.00

Specifications

Antigen Histone H1
Clone r1415-1
Concentration 200μg/mL
Applications Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15940158
View Documents Promotion Details
EnquireBio
3005-MSM7-P1
100 μg
£395.00
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 03-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15930158
View Documents Promotion Details
EnquireBio
3005-MSM7-P0
20 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

Histone H1
200μg/mL
Monoclonal
Mouse
10mM PBS with or without 0.05% BSA and 0.05% azide.
3005
Nuclei of human leukemia biopsy cells
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
∽30kDa
r1415-1
Immunohistochemistry
Unconjugated
RUO
NP_005309
H1
IgG2a κ
Protein A/G Purified
Nuclear
HeLa, A-431, LNCap or Jurkat cells. Breast carcinoma.
Videos
SDS
Documents

Documents

Product Certifications