missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histo-blood group ABO system transferase Antibody (SR1686), Novus Biologicals™
Rabbit Monoclonal Antibody
£240.00 - £325.00
Specifications
| Antigen | Histo-blood group ABO system transferase |
|---|---|
| Clone | SR1686 |
| Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 1:50 |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18695094
|
Novus Biologicals
NBP3-22267-50ul |
50 μL |
£240.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18653785
|
Novus Biologicals
NBP3-22267-100ul |
100 μL |
£325.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Histo-blood group ABO system transferase Monoclonal antibody specifically detects Histo-blood group ABO system transferase in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| Histo-blood group ABO system transferase | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| IgG | |
| Affinity purified |
| SR1686 | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Synthetic peptide from N-terminal | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title