missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histo-blood group ABO system transferase Antibody (SR1686), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22267-100ul
This item is not returnable.
View return policy
Description
Histo-blood group ABO system transferase Monoclonal antibody specifically detects Histo-blood group ABO system transferase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| Histo-blood group ABO system transferase | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Synthetic peptide from N-terminal | |
| 100 μL | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| SR1686 | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction