Learn More
HIST2H2AA4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15302-1-AP-20UL
 View more versions of this product
Product Code. 16894424
  • £135.00 / 20µL
Specifications
HIST2H2AA4
Polyclonal
Unconjugated
HIST2H2AA4
HIST2H2AA4
HIST2H2AA4 Fusion Protein Ag7560
20 μL
Primary
Rat, Human, Mouse
Antibody
IgG
Immunohistochemistry (Paraffin), Western Blot
0.15 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
H2A/R, H2AFO, HIST2H2AA, HIST2H2AA3, HIST2H2AA4, histone cluster 2, H2aa4, Histone H2A type 2 A, Histone H2A.2, Histone H2A/o
Rabbit
Antigen Affinity Chromatography
RUO
723790
-20°C