missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIST2H2AA4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | HIST2H2AA4 |
|---|---|
| Concentration | 0.15 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16894424
|
Proteintech
15302-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16884424
|
Proteintech
15302-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| HIST2H2AA4 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Rat, Human, Mouse | |
| H2A/R, H2AFO, HIST2H2AA, HIST2H2AA3, HIST2H2AA4, histone cluster 2, H2aa4, Histone H2A type 2 A, Histone H2A.2, Histone H2A/o | |
| HIST2H2AA4 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
| 0.15 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| 723790 | |
| HIST2H2AA4 Fusion Protein Ag7560 | |
| Primary | |
| -20°C | |
| HIST2H2AA4 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title