Learn More
Thermo Scientific™ HisPur™ Ni-NTA Spin Purification Kit, 0.2 mL
Shop All Thermo Scientific ProductsDescription
Perform routine affinity purification of His-tagged fusion proteins with Thermo Scientific™ HisPur Ni-NTA Resin, a high-capacity, high-performance nickel-IMAC resin.
HisPur Ni-NTA Resin is available in several package sizes of resin slurry, three sizes of centrifuge-ready columns, complete purification kits, two sizes of FPLC-ready chromatography cartridges, and 96-well filter plates for high through-put needs. With these options, high-yield His-tag protein purification is possible at nearly any scale.
Highlights:
High capacity – bind up to 60mg of 6xHis-tagged protein per milliliter of resin
Versatile – purify proteins using native or denaturing conditions
Compatible – use with Thermo Scientific Cell Lysis Reagents and a variety of buffer additives
Flexible – available in multiple formats including bulk resin, spin columns and chromatography cartridges
Cost-effective – reuse the same batch of resin at least five times
Easy to use – pre-formulated buffers available for kit formats
Available Formats:
- Resin Slurries – crosslinked 6% beaded agarose; 10mL, 100mL, 500mL bottles
- Spin Columns – 0.2mL (microcentrifuge), 1mL, 3mL columns
- Purification Kits – complete kits in all three column sizes
- Chromatography Cartridges – compatible with automated liquid chromatography systems and with syringe processing
- Bulk and Custom – let us know your needs for bulk HisPur Ni-NTA Resin
Includes:
Kits contain HisPur Ni-NTA Columns
Specifications
Specifications
| Capacity (Metric) | 0.2 mL/Column |
| Description | HisPur Ni-NTA Spin Purification Kit, 0.2mL |
| Content And Storage | Sufficient For: Purifying up to 12 mg of His-tagged protein per column • HisPur Ni-NTA Spin Columns, 0.2 mL, 25 columns • Phosphate-buffered Saline (10X), 30 mL • Imidazole (2M), 8 mL • Collection Tubes, 80 tubes • Column Plugs, 25 plugs Store at 4°C. |
| Product Line | HisPur |
| Quantity | 25 Column Kit |
| Target | His-Tagged Protein |
| Type | Spin Column Kit |
| Stationary Phase | Divalent Nickel, Metal Chelate |
| Sufficient For | 12 mg of His-Tagged Protein per Column |
| Format | Kit, Microcentrifuge, Spin Column |
| Show More |
Safety and Handling
- X25 HISPUR NI-NTA PURIFICATION KIT
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Carcinogenicity Category 1A
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1B
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 1 B
- Skin sensitiser Category 1
- Specific target organ toxicity after repeated exposure Category 2
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H350-May cause cancer.
- H360D-May damage the unborn child.
- H360FD-May damage fertility. May damage the unborn child.
- H373-May cause damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Sulfuric acid, nickel(2+) salt, hydrate,sodium chloride, disodium hydrogenorthophosphate, imidazole
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.