Learn More
Thermo Scientific™ HisPur™ Cobalt Purification Kit, 0.2 mL
Gently purify His-tagged recombinant fusion proteins using cobalt (Co2+) immobilized metal chelate affinity (IMAC) beads, spin columns and plates.
Brand: Thermo Scientific™
Code : PN
Additional Details : Weight : 0.10000kg Transport : UN number : 1993 Chem class : 3 Pack group : III
Description
Thermo Scientific™ HisPur Cobalt Resin is an immobilized metal chelate affinity (IMAC) support for specific, fast and gentle His-tagged purification.
Additional Information and Online Ordering »
Highlights:
High purity – obtain more than 10 milligrams of pure His-tagged protein per milliliter of resin without optimizing imidazole washing conditions
Low metal leaching – no metal contamination in eluted histidine-tagged protein sample
Versatility – purify proteins under native or denaturing conditions; compatible with cell lysis reagents and a variety of buffer additives
Flexibility – available as bulk resin or predispensed columns, chromatography columns and 96-well filter plates
Cost effective – resin can be reused several times or discarded after single use
Superior – performs better than other commercially available IMAC Resins
Available Formats:
- Resin Slurries – crosslinked 6% beaded agarose; 10mL, 100mL, 500mL bottles
- Spin Columns – 0.2mL (microcentrifuge), 1mL, 3mL columns
- Purification Kits – complete kits in all three column sizes
- Chromatography Cartridges – compatible with automated liquid chromatography systems and with syringe processing
- 96-well Spin Plates – pre-dispensed 96-well filter plates, compatible with centrifugation and vacuum manifold systems for manual or automated purification. The plates enable fast, consistent well-to-well and plate-to-plate reproducibility for small-scale, high-throughput separations
- Bulk and Custom – let us know your needs for bulk HisPur Cobalt Resin
Includes:
Kits contain HisPur Cobalt Columns, equilibration/wash buffer and elution buffer
Specifications
0.2 mL/Column | |
Sufficient For: Purifying ∼2 mg of His-tagged protein per column • HisPur Cobalt Spin Columns, 0.2 mL, 25 columns • Equilibration/Wash Buffer, 100 mL • Elution Buffer, 50 mL • Collection Tubes, 80 tubes Store at 4°C. |
|
25 Column Kit | |
Protein Purification System | |
2 mg of His-Tagged Protein per column | |
Affinity, Agarose Resin |
HisPur Cobalt Spin Purification Kit, 0.2mL | |
HisPur™ | |
His-Tagged Protein | |
Metal Chelate, Divalent Cobalt | |
Kit, Microcentrifuge, Spin Column |
For Research Use Only. Not for use in diagnostic procedures.