missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HISPPD1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-54642
This item is not returnable.
View return policy
Description
HISPPD1 Polyclonal specifically detects HISPPD1 in Human samples. It is validated for Western Blot.
Specifications
| HISPPD1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| diphosphoinositol pentakisphosphate kinase 2FLJ21506, EC 2.7.4.21, EC 2.7.4.24, HISPPD1histidine acid phosphatase domain containing 1, Histidine acid phosphatase domain-containing protein 1, hsVIP2, inositol heptaphosphate kinase 2, inositol hexakisphosphate and diphosphoinositol-pentakisphosphate kinase 2, InsP6 and PP-IP5 kinase 2, KIAA0433FLJ23463, VIP1 homolog 2, VIP2IP7K2 | |
| Rabbit | |
| 138 kDa | |
| 100 μL | |
| Protein Phosphatase | |
| 23262 | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| O43314 | |
| PPIP5K2 | |
| Synthetic peptides corresponding to HISPPD1(histidine acid phosphatase domain containing 1) The peptide sequence was selected from the middle region of HISPPD1. Peptide sequence SLSSCQQRVKARLHEILQKDRDFTAEDYEKLTPSGSISLIKSMHLIKNPV. | |
| Affinity purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction