missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiScreen™ Capto™ MMC Multimodal Prepacked Column
Shop All Cytiva Products
Click to view available options
Quantity:
1 x 4.7mL
Unit Size:
Each
Description
- With innovative salt-tolerant ligand
- Weak multimodal cation exchanger with a different selectivity compared with traditional ion exchangers
- Highly cross-linked agarose matrix
- Ready-to-use columns for convenient process development optimization
- Bed height of 10cm HiScreen columns is designed to allow method optimization and parameter screening
- Can be easily connected in series to achieve 20cm bed height
- Small bed volume gives fast results and minimal sample/buffer consumption
- Reproducible results, scalable to BioProcess columns packed with the same media using the same linear fluid velocity
Specifications
Specifications
| Buffer | 1M Acetic Acid, 1M NaOH, 8M Urea, 6M Guanidine Hydrochloride and 70% Ethanol |
| Product Type | HiScreen Capto MMC column |
| Bed Volume | 4.7mL |
| Diameter (English) Inner | 0.3 in. |
| Diameter (Metric) Inner | 7.7mm |
| Quantity | 1 x 4.7mL |
| Height (English) | 3.93 in. |
| Height (Metric) | 100mm |
| Flow Rate | <600cm/hr. |
| Material | Polypropylene |
| Show More |
Safety and Handling
Product Identifier
- HiScreen Capto MMC
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction