missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiScreen Capto™ Core 700 Columns

Product Code. 12663850 Shop All Cytiva Products
Click to view available options
Quantity:
1 x 4.7mL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12663850

Brand: Cytiva 17548115

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Capto Core 700 is designed for intermediate purification and polishing of viruses and other large biomolecules.

The novel core bead technology and multimodal, octylamine ligand give Capto Core 700 dual functionality, size exclusion, and binding chromatography in one chromatography medium (resin).

  • For intermediate purification and polishing of viruses and other large biomolecules (Mr > 700 000) in flowthrough mode.
  • Significantly improved productivity and higher flow rates compared with gel filtration methods.
  • Straightforward optimization due to flowthrough chromatography and robust performance.
  • Convenient small-scale purification, process development, and scale-up using prepacked HiTrap and HiScreen columns.
  • Capto Core 700 is composed of a ligand-activated core and inactive shell.
  • The inactive shell excludes large molecules (cut off ∼ Mr 700 000 [700 kDa]) from entering the core through the pores of the shell.
  • The core of each bead is functionalized with ligands that are both hydrophobic and positively charged, resulting in a highly efficient multimodal binding of various contaminants small enough to enter the core.
  • The multimodal ligands ensure strong binding with most impurities over a wide range of pH and salt concentrations.
  • Bound impurities are removed from the beads by cleaning-in-place (CIP) procedures using NaOH and in most cases, a solvent.

TRUSTED_SUSTAINABILITY

Specifications

Buffer 1M NaOH, 6M Guanidine Hydrochloride, 30% Isopropanol and 70% Ethanol
Product Type HiScreen Capto Core 700 multimodal column
Bed Volume 4.7mL
Diameter (English) Inner 0.30 in.
Diameter (Metric) Inner 7.7mm
Quantity 1 x 4.7mL
Flow Rate <500cm/hr.
Material Polypropylene
Ligand Type Octylamine
Max. Pressure 8 bar
pH Range 2 to 14
Particle Size 90μm
Support Type Highly Cross-linked Agarose
Content And Storage 4° to 30°C
Column Type Multimodal Column
For Use With (Application) Intermediate purification and polishing of viruses and other large biomolecules
Show More Show Less
compliance-icons
Product Identifier
  • HiScreen Capto
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P273-Avoid release to the environment.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Ethanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.