missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiScreen™ Capto™ adhere Multimodal Prepacked Column

Product Code. 10201180 Shop All Cytiva Products
Click to view available options
Quantity:
1 x 4.7mL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10201180

Brand: Cytiva 28926981

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For intermediate purification and polishing of monoclonal antibodies after the protein A capture step

  • Strong multimodal anion exchanger with N-benzyl-n-methyl ethanolamine ligand
  • Offers alternative selectivity compared to traditional anion exchangers
  • In flowthrough mode, removal of leached protein A, aggregates, host cell proteins, nucleic acids and viruses from MAbs is performed
  • Antibodies pass directly through the column while the contaminants remain bound
  • Highly cross-linked agarose matrix
  • Ready-to-use columns for convenient process development optimization
  • Bed height of 10cm is designed to allow method optimization and parameter screening
  • Easily connected in series to achieve 20cm bed height

TRUSTED_SUSTAINABILITY

Specifications

Buffer 1M Acetic Acid, 1M NaOH
Product Type HiScreen Capto adhere column
Bed Volume 4.7mL
Diameter (English) Inner 0.3 in.
Diameter (Metric) Inner 7.7mm
Quantity 1 x 4.7mL
Height (English) 3.93 in.
Height (Metric) 100mm
Flow Rate <600cm/hr.
Material Polypropylene
Ligand Type N-benzyl-n-methyl Ethanolamine
Max. Pressure 3 bar
pH Range 3 to 12
Particle Size 75μm
Support Type Highly Cross-linked Agarose
Content And Storage 4° to 30°C
Column Type Multimodal Column
For Use With (Application) Optimization and parameter screening
Show More Show Less
compliance-icons
Product Identifier
  • HiScreen Capto adhere
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.