missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiScreen™ Blue, Ni and DEAE Sepharose™ Prepacked Fast Flow Columns

Product Code. 11370322 Shop All Cytiva Products
Change view
Click to view available options
Ligand Type:
Blue Sepharose
DEAE Sepharose
Ni Sepharose
Unit Size:
Each
Product Code. Ligand Type unitSize
11370322 DEAE Sepharose Each
12305683 Ni Sepharose Each
11603234 Blue Sepharose Each
3 options
This item is not returnable. View return policy

Product Code. 11370322

Brand: Cytiva 28978245

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for method optimization and parameter screening

  • Based on 6% highly cross-linked agarose with an average bead size of 90μm
  • Processed flow properties and high loading capacities
  • Bed height of 10cm is designed to allow method optimization and parameter screening
  • Easily connected in series to achieve 20cm bed height
  • Small bed volume gives fast results and minimal sample/buffer consumption
  • Combines high capacity with high flow velocity and low backpressure to reduce process cycle times and increase productivity
  • Reproducible results, scalable to BioProcess™ columns packed with the same chromatography media using the same linear fluid velocity

  • Bed volume: 4.7mL
  • Bed dimension: 7.7 × 100mm
  • Matrix: 6% cross-linked agarose beads
  • Average particle size: 90μm
  • Flow velocity: <600cm/hr or <450cm/hr
  • Material (Column hardware and column tube): polypropylene
  • Column I.D.: 7.7mm

Blue Sepharose

  • Prepacked with BioProcess chromatography medium Blue Sepharose 6 Fast Flow
  • Purification of albumin, enzymes (including NAD+ and NADP+), coagulation factors, interferons and related proteins
Ni Sepharose
  • Prepacked with Nickel Sepharose 6 Fast Flow
  • Purification of histidine-tagged proteins with Immobilized Metal ion Affinity Chromatography (IMAC)
DEAE Sepharose
  • Prepacked with DEAE Sepharose Fast Flow
  • Weak anion exchanger (diethylaminoethyl group) BioProcess™ medium

TRUSTED_SUSTAINABILITY

Specifications

Buffer 1M NaOH, 8M Urea, 8M Guanidine Hydrochloride, 70% Ethanol
Product Type HiScreen DEAE Sepharose FF column
Bed Volume 4.7 mL
Diameter (English) Inner 0.3 in.
Diameter (Metric) Inner 7.7 mm
Quantity 1 x 4.7mL
Height (English) 3.93 in.
Height (Metric) 100mm
Flow Rate <450 cm/hr.
Material Polypropylene
Ligand Type DEAE Sepharose
Max. Pressure 1.5 bar
pH Range 2 to 12
Particle Size 45 to 165μm
Support Type 6% Cross-linked Agarose
Content And Storage 4° to 30°C
Column Type Ion Exchange Column
For Use With (Application) Convenient process development optimization
Show More Show Less
compliance-icons
Product Identifier
  • HiScreen DEAE FF
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.