missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiScale™ Chromatography Columns

Withstand pressures of up to 20 bar (290 psi) and high flow rates. Cytiva Life Sciences™ HiScale™ Chromatography Columns are designed to be compatible with modern BioProcess chromatography media such as MabSelect™ and Capto™.

Brand:  Cytiva 28-9645-13

Code : GX

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 11348390

  • £1074.00 / Each
Estimated Shipment: 06-06-2024
to see stock.
Add to basket

Description

Description

  • Empty columns with high pressure stability for packing with modern BioProcess media for process development and preparative chromatography.
  • Provides axial compression of the gel bed and a nonrotating plunger mechanism that expands packing capabilities.
  • Adapter QuickLock mechanism, column measurement scale, and ergonomic design, facilitate column handling and increase process control.
  • Dual adapters, offering high flexibility and dynamic packing heights.
  • Capability to rinse entire adapter shaft after packing, thereby minimizing risk of bacterial growth after prolonged use.
  • Based on the well-established line of XK columns and compatible with all methods currently applicable to XK columns.
Specifications

Specifications

HiScale Chromatography Column
26mm
Column tube, outer protection tube, two adapters, PEEK tubing, and unions at both ends for easy connection to valves, pumps, and monitors
20 bar
Empty HPLC Column
212mL
400mm
Borosilicate glass
Polypropylene (reinforced glassfibre)/Polyamide Net
Process development and preparative chromatography
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers