missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Single-use spin columns prepacked with Ni Sepharose High Performance Medium that is precharged with nickel ions; negligible ion leakage
- One run in standard microcentrifuge takes approximatedly 10 min.
- Yields up to 750μg purified protein per column
- Allows direct purification of both clarified and unclarifed cell lysates
- Also useful for screening large numbers of small-scale bacterial lysates
- Kit option contains buffer, allowing fast and convenient purifications without time-consuming buffer preparation step
Medium Characteristics:
- Compatible with a wide range of reducing agents, detergents, denaturants and other additives
- Stable (long-term) in the pH range 3-12
- Average particle size: 34μm
Order Info
Kit (28-9321-71) contains 50 columns and one pack of His Buffer Kit (2 x 100mL phosphate 8X stock solution, pH 7.4, and 100mL highly pure 2M imidazole, pH 7.4)
Specifications
Specifications
| Bed Volume | 100 μL |
| pH Range | 3 to 12 |
| Support Type | 6% Highly Cross-linked Agarose |
| Content And Storage | 4°C to 30°C |
| Quantity | 1 Pack His SpinTrap Spin Columns (50 x 100 μL) |
| Type | Spin Column |
| Sample Volume | 600 μl |
Safety and Handling
Signal Word
- Warning
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P501-Dispose of contents/container to .
- MIXTURE LIST-Contain: nicke, reaction massof:5-chloro-2-methyl-4-isothiazolin-3-one and 2-methyl-2Hisothiazol-3-one
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?