missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Samples that usually cause stripping of metal ions can therefore be added to the medium with retained protein-binding capacity and without pretreatment.
- His Mag Sepharose excel is a magnetic IMAC medium precharged with nickel ions that are very strongly bound to a chelating ligand.
- The medium enables an efficient and simplified workflow for capture, purification, and reproducible screening of histidine-tagged proteins.
- Simplify small-scale purification and expression screening of histidine-tagged proteins secreted in eukaryotic cell culture.
- Purify histidine-tagged proteins directly from samples that contain metal ion-stripping agents and save time and effort.
- Increase target protein yield and decrease degradation through reduced and simplified sample handling.
- Screen histidine-tagged proteins in parallel with high reproducibility and reliable results.
- Capture histidine-tagged protein from different sample volumes; from low to high milliliter scale.
- Simplify handling using visible and dense Sepharose-based magnetic beads.
- Proteins are expressed and secreted into cell culture supernatants from eukaryotic cells such as insect cells or Chinese Hamster Ovary (CHO) cells.
- The high density of the beads allows rapid capture by magnetic devices while the visibility of the beads ensures reliable collection of the bound histidine-tagged proteins during the purification procedure.
- The base matrix is based on Sepharose medium with magnetite incorporated.
- His Mag Sepharose is available as 10% medium slurry in three different pack sizes (2 x 1ml, 5 x 1ml, and 10 x 1ml) and can be used together with different test tubes and magnetic racks.
Specifications
Specifications
| pH Range | 2 to 12 |
| Support Type | Highly cross-linked spherical Agarose including Magnetite |
| Content And Storage | 4°C to 30°C |
| Particle Size | 63 μm |
| Quantity | 10 x 1 mL |
| For Use With (Application) | Designed to simplify small-scale purification and screening of His-tagged Proteins secreted in eukaryotic cell culture, used for functional and structural studies research in academia, institutes and discovery Pharma |
| Type | Chromatography Media |
Safety and Handling
Signal Word
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- EUH208-Contains Nickel. May produce an allergic reaction.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : ethanol
- UNKNOWN AQUATIC-Contains 50% of components with unknown hazards to the aquatic environment.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?