missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HIPK3 (YAK1) Recombinant Human Protein
HIPK3 is a serine/threonine kinase.
Brand: Invitrogen™ PV4324
Code : NEW
Additional Details : Weight : 0.00900kg
Description
HIPK3 is involved in signal transduction, cellular communication, and multidrug resistance of tumors and plays a role in the phosphorylation of FADD.
Specifications
NP_005725.2 | |
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol | |
1mg | |
RUO - research use only | |
HIPK3 (YAK1) | |
Human | |
His Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
49.7kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Catalytic | |
DYRK Kinase Family | |
Recombinant | |
Baculovirus |