missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HIPK3 (YAK1) Recombinant Human Protein
HIPK3 is a serine/threonine kinase.
£1768.00 - £11240.00
Specifications
Accession Number | NP_005725.2 |
---|---|
For Use With (Application) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Formulation | 50mM Tris-HCl pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol |
Molecular Weight (g/mol) | 49.7kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15478613
|
Invitrogen™
PR7715A |
100μg |
£1768.00
100µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10577533
|
Invitrogen™
PV4324 |
1mg |
£11240.00
1mg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
HIPK3 is involved in signal transduction, cellular communication, and multidrug resistance of tumors and plays a role in the phosphorylation of FADD.
Specifications
NP_005725.2 | |
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Catalytic | |
DYRK Kinase Family | |
Recombinant | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
49.7kDa | |
RUO - research use only | |
HIPK3 (YAK1) | |
Human | |
His Tag | |
Frozen |