missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIPK3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£167.00 - £326.00
Specifications
| Antigen | HIPK3 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 0.5-2 ug/ml, Immunocytochemistry/Immunofluorescence 1:200, Immunohistochemistry-Paraffin 5-10 ug/ml, Immunocytochemistry 2-10 ug/ml |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18190962
|
Novus Biologicals
NBP2-24521SS |
0.025 mg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18727773
|
Novus Biologicals
NBP2-24521 |
0.1 mg |
£326.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HIPK3 Polyclonal specifically detects HIPK3 in Human, Mouse, Rat, Canine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| HIPK3 | |
| Western Blot 0.5-2 ug/ml, Immunocytochemistry/Immunofluorescence 1:200, Immunohistochemistry-Paraffin 5-10 ug/ml, Immunocytochemistry 2-10 ug/ml | |
| Polyclonal | |
| Rabbit | |
| Protein Kinase | |
| PBS with 0.05% Sodium Azide | |
| Androgen receptor-interacting nuclear protein kinase, DYRK6Fas-interacting serine/threonine-protein kinase, EC 2.7.11.1, FIST, FIST3ANPK, homeodomain interacting protein kinase 3, homeodomain-interacting protein kinase 3, PKYHomolog of protein kinase YAK1, YAK1 | |
| HIPK3 | |
| IgG | |
| Affinity Purified | |
| The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a and CRA_c) and chimpanzee, 94% homologous in orangutan and rhesus monkey, 83% homologous in horse and mouse, 77% homologous in cow and 47% homologous in rat. |
| 1 mg/mL | |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat, Canine, Primate | |
| Q9H422 | |
| 10114 | |
| A portion of amino acids 850-900 of human HIPK3 was used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title