missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIPK3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-24521SS
This item is not returnable.
View return policy
Description
HIPK3 Polyclonal specifically detects HIPK3 in Human, Mouse, Rat, Canine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| HIPK3 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| Androgen receptor-interacting nuclear protein kinase, DYRK6Fas-interacting serine/threonine-protein kinase, EC 2.7.11.1, FIST, FIST3ANPK, homeodomain interacting protein kinase 3, homeodomain-interacting protein kinase 3, PKYHomolog of protein kinase YAK1, YAK1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a and CRA_c) and chimpanzee, 94% homologous in orangutan and rhesus monkey, 83% homologous in horse and mouse, 77% homologous in cow and 47% homologous in rat. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 0.5-2 ug/ml, Immunocytochemistry/Immunofluorescence 1:200, Immunohistochemistry-Paraffin 5-10 ug/ml, Immunocytochemistry 2-10 ug/ml | |
| Q9H422 | |
| HIPK3 | |
| A portion of amino acids 850-900 of human HIPK3 was used as the immunogen. | |
| 0.025 mg | |
| Protein Kinase | |
| 10114 | |
| Human, Mouse, Rat, Canine, Primate | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction