missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HIPK2 Recombinant Human Protein
HIPK2 is a nuclear serine/threonine kinase.
Brand: Invitrogen™ PR9004A
Code : NEW
Additional Details : Weight : 0.01000kg
Description
HIPK2 is linked to transcriptional regulation and apoptosis.Specifications
NP_073577.3 | |
73kDa | |
Kinase proteins are shipped on dry ice and should be stored at -80°C. | |
Catalytic | |
DYRK Kinase Family | |
Recombinant | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
100μg | |
RUO - research use only | |
HIPK2 | |
Human | |
GST Tag | |
Frozen |