missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HIPK2 Recombinant Human Protein
HIPK2 is a nuclear serine/threonine kinase.
£434.00 - £6900.00
Specifications
Accession Number | NP_073577.3 |
---|---|
Molecular Weight (g/mol) | 73kDa |
Storage Requirements | Kinase proteins are shipped on dry ice and should be stored at -80°C. |
Gene Alias | HIPK2 |
KinaseFamily | DYRK Kinase Family |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10175653
|
Invitrogen™
PV5275 |
10μg |
£434.00
Each |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15458703
|
Invitrogen™
PR9004A |
100μg |
£1604.00
100µg |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15458783
|
Invitrogen™
PV5276 |
1mg |
£6900.00
1mg |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
HIPK2 is linked to transcriptional regulation and apoptosis.Specifications
NP_073577.3 | |
Kinase proteins are shipped on dry ice and should be stored at -80°C. | |
DYRK Kinase Family | |
Recombinant | |
Baculovirus |
73kDa | |
HIPK2 | |
Human | |
GST Tag |